Learn More
ABclonal Technology GBX2 Rabbit pAb
SDP
Catalog No. 5000108906
Encompass_Preferred
Enables sequence-specific double-stranded DNA binding activity. Predicted to be involved in regulation of nervous system development and regulation of transcription by RNA polymerase II. Predicted to act upstream of or within several processes, including branching involved in blood vessel morphogenesis, nervous system development, and neural crest cell migration. Predicted to be part of chromatin. Predicted to be active in nucleus.
Catalog No. 50-001-08906 Supplier ABclonal Technology Supplier No. A14733500UL
Enables sequence-specific double-stranded DNA binding activity. Predicted to be involved in regulation of nervous system development and regulation of transcription by RNA polymerase II. Predicted to act upstream of or within several processes, including branching involved in blood vessel morphogenesis, nervous system development, and neural crest cell migration. Predicted to be part of chromatin. Predicted to be active in nucleus.