Learn More
Aviva Systems Biology Corp GC Antibody - N-terminal region (ARP61150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GC. It was validated on Western Blot. . Alternate names: DBP, DBP/GC, GRD3, VDBG, VDBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-272