Learn More
Aviva Systems Biology Corp GC Antibody - N-terminal region (ARP61151_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61151_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GC. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GC. Alternate names: DBP, DBP/GC, GRD3, VDBG, VDBPApplicationWB. Species reactivity: Horse, Human, Pig, Rat
Catalog No. 50-287-273