Learn More
Agilent Technologies GC column: DB-1701 30m x 0.53mm x 1um
Catalog No. NC2742316
Encompass
J&W DB-1701 GC Column, 30 m, 0.53 mm, 1.00 µm, 7 inch cage. This is the most common GC column format compatible with Agilent 5890, 6890, 7820, 7890, 8860, and 8890 series GC systems and all non-Agilent GC systems with a similarly sized oven.
Catalog No. NC2742316 Supplier Agilent Technologies Supplier No. 1250732
J&W DB-1701 GC Column, 30 m, 0.53 mm, 1.00 µm, 7 inch cage. This is the most common GC column format compatible with Agilent 5890, 6890, 7820, 7890, 8860, and 8890 series GC systems and all non-Agilent GC systems with a similarly sized oven.