Learn More
Azzota Corp Gc Column, Gc Mid-Polar, Usp Code: G46; Gsbp-1701Ms, 30M X 0.25mm X 0.25um.
Catalog No. 501953944
Encompass_Preferred
Gc Column, Gc Mid-Polar, Usp Code: G46; Gsbp-1701Ms, 30M X 0.25Mm X 0.25um.
Catalog No. 50-195-3944 Supplier Azzota Corp Supplier No. 67253002
Gc Column, Gc Mid-Polar, Usp Code: G46; Gsbp-1701Ms, 30M X 0.25Mm X 0.25um.