Learn More
Azzota Corp Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1, 30M X 0.25mm X 0.25um. Equivalent To Hp-1, Db-1, Cp-Sil 5Cb, Vf-1, Rtx-1, Rxi-1, Sp-1 Zb-1, Bp-1.
Catalog No. 501953923
Encompass_Preferred
Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-1, Db-1, Cp-Sil 5Cb, Vf-1, Rtx-1, Rxi-1, Sp-1 Zb-1, Bp-1.
Catalog No. 50-195-3923 Supplier Azzota Corp Supplier No. 01253002
Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-1, Db-1, Cp-Sil 5Cb, Vf-1, Rtx-1, Rxi-1, Sp-1 Zb-1, Bp-1.