Learn More
Azzota Corp Gc Column, Gc Polar-Peg, Usp Code: G16; Gsbp-Wax-Aq, 30M X 0.25mm X 0.25um.
Catalog No. 501953937
Encompass_Preferred
Gc Column, Gc Polar-Peg, Usp Code: G16; Gsbp-Wax-Aq, 30M X 0.25Mm X 0.25um. Equivalent To .
Catalog No. 50-195-3937 Supplier Azzota Corp Supplier No. 26253002
Gc Column, Gc Polar-Peg, Usp Code: G16; Gsbp-Wax-Aq, 30M X 0.25Mm X 0.25um. Equivalent To .