Learn More
Azzota Corp Gc Column, Gc Non-Polar, Usp Code: G27; Gsbp-5Msinert, 30M X 0.25mm X 0.25um. Equivalent To Hp-5Ms, Db-5Ms, Cp-Sil 8Cb Ms, Vf-5Ms, Rtx-5Ms, Rxi-5Ms, Sp-5Ms, Zb-5Ms.
Catalog No. 501953929
Encompass_Preferred
Gc Column, Gc Non-Polar, Usp Code: G27; Gsbp-5Msinert, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-5Ms, Db-5Ms, Cp-Sil 8Cb Ms, Vf-5Ms, Rtx-5Ms, Rxi-5Ms, Sp-5Ms, Zb-5Ms.
Catalog No. 50-195-3929 Supplier Azzota Corp Supplier No. 15253002UI
Gc Column, Gc Non-Polar, Usp Code: G27; Gsbp-5Msinert, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-5Ms, Db-5Ms, Cp-Sil 8Cb Ms, Vf-5Ms, Rtx-5Ms, Rxi-5Ms, Sp-5Ms, Zb-5Ms.