Learn More
Azzota Corp Gc Column, Gc Polar-Peg, Usp Code: G16; Gsbp-Carbowax 20M, 30M X 0.25mm X 0.25um. Equivalent To Hp-20M, Db-Cam, Cp-Sil 57Wax, .
Catalog No. 501953935
Encompass_Preferred
Gc Column, Gc Polar-Peg, Usp Code: G16; Gsbp-Carbowax 20M, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-20M, Db-Cam, Cp-Sil 57Wax, .
Catalog No. 50-195-3935 Supplier Azzota Corp Supplier No. 22253002
Gc Column, Gc Polar-Peg, Usp Code: G16; Gsbp-Carbowax 20M, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-20M, Db-Cam, Cp-Sil 57Wax, .