Learn More
Azzota Corp Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1Ms, 30M X 0.25mm X 0.25um. Equivalent To Hp-1Ms, Db-1Ms, Cp-Sil 5Cb Ms, Vf-1Ms, Rtx-1Ms, Rxi-1Ms.
Catalog No. 501953927
Encompass_Preferred
Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1Ms, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-1Ms, Db-1Ms, Cp-Sil 5Cb Ms, Vf-1Ms, Rtx-1Ms, Rxi-1Ms.
Catalog No. 50-195-3927 Supplier Azzota Corp Supplier No. 11253002
Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1Ms, 30M X 0.25Mm X 0.25Um. Equivalent To Hp-1Ms, Db-1Ms, Cp-Sil 5Cb Ms, Vf-1Ms, Rtx-1Ms, Rxi-1Ms.