Learn More
Azzota Corp Gc Column, Gc Mid-Polar, Usp Code: G3; Gsbp-50+Ms, 30M X 0.25mm X 0.25um. Equivalent To Db-17, Db-17Ms, Hp-50+, Cp-Sil 24Cb, Vf-17Ms, Rxt-50, Rtx-17.
Catalog No. 501953938
Encompass_Preferred
Gc Column, Gc Mid-Polar, Usp Code: G3; Gsbp-50+Ms, 30M X 0.25Mm X 0.25Um. Equivalent To Db-17, Db-17Ms, Hp-50+, Cp-Sil 24Cb, Vf-17Ms, Rxt-50, Rtx-17.
Catalog No. 50-195-3938 Supplier Azzota Corp Supplier No. 50253002
Gc Column, Gc Mid-Polar, Usp Code: G3; Gsbp-50+Ms, 30M X 0.25Mm X 0.25Um. Equivalent To Db-17, Db-17Ms, Hp-50+, Cp-Sil 24Cb, Vf-17Ms, Rxt-50, Rtx-17.