Learn More
Azzota Corp Gc Column, Gc Non-Polar, Usp Code: G27; Gsbp-5, 30M X 0.32mm X 0.25um. Equivalent To Hp-5, Db-5, Cp-Sil 8Cb, Vf-5Ms, Rtx-5, Rxi-5, Sp-, Zb-5, Bp-5.
Catalog No. 501953926
Encompass_Preferred
Gc Column, Gc Non-Polar, Usp Code: G27; Gsbp-5, 30M X 0.32Mm X 0.25Um. Equivalent To Hp-5, Db-5, Cp-Sil 8Cb, Vf-5Ms, Rtx-5, Rxi-5, Sp-, Zb-5, Bp-5.
Catalog No. 50-195-3926 Supplier Azzota Corp Supplier No. 05323002
Gc Column, Gc Non-Polar, Usp Code: G27; Gsbp-5, 30M X 0.32Mm X 0.25Um. Equivalent To Hp-5, Db-5, Cp-Sil 8Cb, Vf-5Ms, Rtx-5, Rxi-5, Sp-, Zb-5, Bp-5.