Learn More
Azzota Corp Gc Column, Gc Mid-Polar, Usp Code: G43; Gsbp-624, 30M X 0.32mm X 1.80um. Equivalent To Db-624, Db-Vrx, Rtx-624, Rtx-Vms, Db 502.2, Voccol
Catalog No. 501953942
Encompass_Preferred
Gc Column, Gc Mid-Polar, Usp Code: G43; Gsbp-624, 30M X 0.32Mm X 1.80Um. Equivalent To Db-624, Db-Vrx, Rtx-624, Rtx-Vms, Db 502.2, Voccol.
Catalog No. 50-195-3942 Supplier Azzota Corp Supplier No. 62323018
Gc Column, Gc Mid-Polar, Usp Code: G43; Gsbp-624, 30M X 0.32Mm X 1.80Um. Equivalent To Db-624, Db-Vrx, Rtx-624, Rtx-Vms, Db 502.2, Voccol.