Learn More
Azzota Corp Gc Column, Gc Polar-Peg, Usp Code: G35; Gsbp-Ffap, 30M X 0.53mm X 1.00um. Equivalent To Hp-Ffap, Db-Ffap, Rxt-Stabilwa-Da, Cp-Ffap.
Catalog No. 501953934
Encompass_Preferred
Gc Column, Gc Polar-Peg, Usp Code: G35; Gsbp-Ffap, 30M X 0.53Mm X 1.00Um. Equivalent To Hp-Ffap, Db-Ffap, Rxt-Stabilwa-Da, Cp-Ffap.
Catalog No. 50-195-3934 Supplier Azzota Corp Supplier No. 21533010
Gc Column, Gc Polar-Peg, Usp Code: G35; Gsbp-Ffap, 30M X 0.53Mm X 1.00Um. Equivalent To Hp-Ffap, Db-Ffap, Rxt-Stabilwa-Da, Cp-Ffap.