Learn More
Azzota Corp Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1, 30M X 0.53mm X 1.0um. Equivalent To Hp-1, Db-1, Cp-Sil 5Cb, Vf-1, Rtx-1, Rxi-1, Sp-1 Zb-1, Bp-1.
Catalog No. 501953924
Encompass_Preferred
Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1, 30M X 0.53Mm X 1.0Um. Equivalent To Hp-1, Db-1, Cp-Sil 5Cb, Vf-1, Rtx-1, Rxi-1, Sp-1 Zb-1, Bp-1.
Catalog No. 50-195-3924 Supplier Azzota Corp Supplier No. 01533010
Gc Column, Gc Non-Polar, Usp Code: G2; Gsbp-1, 30M X 0.53Mm X 1.0Um. Equivalent To Hp-1, Db-1, Cp-Sil 5Cb, Vf-1, Rtx-1, Rxi-1, Sp-1 Zb-1, Bp-1.