Learn More
Azzota Corp Gc Column, Gc Mid-Polar, Usp Code: G43; Gsbp-624, 30M X 0.53mm X 3.00um. Equivalent To Db-624, Db-Vrx, Rtx-624, Rtx-Vms, Db 502.2, Voccol.
Catalog No. 501953943
Encompass_Preferred
Gc Column, Gc Mid-Polar, Usp Code: G43; Gsbp-624, 30M X 0.53Mm X 3.00Um. Equivalent To Db-624, Db-Vrx, Rtx-624, Rtx-Vms, Db 502.2, Voccol.
Catalog No. 50-195-3943 Supplier Azzota Corp Supplier No. 62533030
Gc Column, Gc Mid-Polar, Usp Code: G43; Gsbp-624, 30M X 0.53Mm X 3.00Um. Equivalent To Db-624, Db-Vrx, Rtx-624, Rtx-Vms, Db 502.2, Voccol.