Learn More
Waters Corp GC μECD ALS Qualification Standards Kit, Liquid, 10 mL
Catalog No. 502392054 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GC μECD ALS Qualification Standards Kit is for system assessment of Gas Chromatography (GC) systems with a micro Electron Capture Detector (μECD) and liquid autosampler. It provides standards to verify system reproducibility and conduct functional tests, ensuring optimal GC-MS workflow calibration and accurate compound quantitation.
  • Designed for GC systems with a μECD detector and liquid autosampler.
  • Contains standards for verifying system reproducibility and other functional tests.
  • Formulated to meet or exceed the needs of gas chromatography systems.
  • Helps calibrate the GC-MS workflow.
  • Enables verification of system performance for accurate compound quantitation.
Catalog No. 50-239-2054 Supplier Waters Corp Supplier No. 186007653
This GC μECD ALS Qualification Standards Kit is for system assessment of Gas Chromatography (GC) systems with a micro Electron Capture Detector (μECD) and liquid autosampler. It provides standards to verify system reproducibility and conduct functional tests, ensuring optimal GC-MS workflow calibration and accurate compound quantitation.
  • Designed for GC systems with a μECD detector and liquid autosampler.
  • Contains standards for verifying system reproducibility and other functional tests.
  • Formulated to meet or exceed the needs of gas chromatography systems.
  • Helps calibrate the GC-MS workflow.
  • Enables verification of system performance for accurate compound quantitation.