Learn More
Waters Corp Qualification Standards Kit, GC, μECD, Headspace
Catalog No. 502390577 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GC standards kit is designed for system assessment of GC systems that include a μECD detector and a headspace autosampler. The kit provides standards suitable for verifying system reproducibility and conducting other functional tests.
  • Designed for system assessment of gas chromatograph systems.
  • Includes standards for verifying system reproducibility.
  • Suitable for conducting other functional tests.
  • Essential for ensuring proper operation of the μECD detector.
  • Formulated for detecting trace compounds in samples.
  • For use with headspace autosamplers.
Catalog No. 50-239-0577 Supplier Waters Corp Supplier No. 186007584
This GC standards kit is designed for system assessment of GC systems that include a μECD detector and a headspace autosampler. The kit provides standards suitable for verifying system reproducibility and conducting other functional tests.
  • Designed for system assessment of gas chromatograph systems.
  • Includes standards for verifying system reproducibility.
  • Suitable for conducting other functional tests.
  • Essential for ensuring proper operation of the μECD detector.
  • Formulated for detecting trace compounds in samples.
  • For use with headspace autosamplers.