Learn More
Waters Corp Qualification Standards Kit, Liquid, 20 mL, 12 Months Shelf Life
Catalog No. 502391282 Shop All Waters Corporation Products
Encompass_Preferred
This GC standards kit assesses GC systems with Flame Ionization Detector (FID) and headspace autosampler. It provides standards for verifying system reproducibility and functional tests, aiding qualification. Kits include Material Safety Data Sheet and Certificate of Analysis. Contains hazardous materials, requiring special handling.
  • Reference standards
  • System assessment and qualification for GC-FID systems with headspace autosamplers
  • Standards for verifying system reproducibility and functional tests
  • Liquid formulation
  • FID detection method
  • GC separation mode
  • GC-FID system type
  • 12 months shelf life
  • Room temperature storage condition
  • Ambient shipping condition
  • Volume: 20 mL
  • 1 unit per package
  • Hazardous material; special freight handling required
  • Material safety data sheet and certificate of analysis included
Catalog No. 50-239-1282 Supplier Waters Corp Supplier No. 186007585
This GC standards kit assesses GC systems with Flame Ionization Detector (FID) and headspace autosampler. It provides standards for verifying system reproducibility and functional tests, aiding qualification. Kits include Material Safety Data Sheet and Certificate of Analysis. Contains hazardous materials, requiring special handling.
  • Reference standards
  • System assessment and qualification for GC-FID systems with headspace autosamplers
  • Standards for verifying system reproducibility and functional tests
  • Liquid formulation
  • FID detection method
  • GC separation mode
  • GC-FID system type
  • 12 months shelf life
  • Room temperature storage condition
  • Ambient shipping condition
  • Volume: 20 mL
  • 1 unit per package
  • Hazardous material; special freight handling required
  • Material safety data sheet and certificate of analysis included