Learn More
Athens Research And Technology Inc GC GLOBULIN HU 1MG
Supplier:  Athens Research And Technology Inc 16160703071MG
Encompass_Preferred
GC Globulin, Human Plasma (Vitamin D Binding Protein): 1 MG; Lyophilized from 20 mM Na phosphate, pH 7.4, with 150 mM NaCl; MW 56,000; Purity: > =95% by SDS-PAGE.
Catalog No. 50-489-741