Learn More
AB Vector LLC GC (Green Control), 1 mL
Catalog No. NC1600074
Encompass
Positive control baculovirus in experiments where GFP fluorescence is used to monitor virus propagation, optimize cultivation conditions, or titrate of virus stocks.
Catalog No. NC1600074 Supplier AB Vector LLC Supplier No. C14
Positive control baculovirus in experiments where GFP fluorescence is used to monitor virus propagation, optimize cultivation conditions, or titrate of virus stocks.