Learn More
Waters Corp Waters Corporation GC Headspace Qualification Standards Kit, 9 Ampules, 20 mL, Acetone Solvent
Catalog No. 50201691 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Gas Chromatograph standards kit is designed for system assessment. It contains 9 ampules, each with 20 mL of standard solutions, including 3 ampules of a Blank (Acetone) solution and 6 ampules containing 0.1% o-Xylene and 0.02% 1,2,4-Trichlorobenzene, all prepared in Acetone (v/v).
  • Used for system assessment in gas chromatography.
  • Hazardous material, requiring special freight handling.
Catalog No. 50-201-691 Supplier Waters Corp Supplier No. 700005095
This Gas Chromatograph standards kit is designed for system assessment. It contains 9 ampules, each with 20 mL of standard solutions, including 3 ampules of a Blank (Acetone) solution and 6 ampules containing 0.1% o-Xylene and 0.02% 1,2,4-Trichlorobenzene, all prepared in Acetone (v/v).
  • Used for system assessment in gas chromatography.
  • Hazardous material, requiring special freight handling.