Learn More
Phenomenex Inc GC Liner for HP, Split, Straight
Catalog No. 502395241 Shop All Phenomenex Products
Encompass_Preferred
GC Liner for HP, Split, Straight 4mm ID x 78.5mm L x 6.3mm OD
Catalog No. 50-239-5241 Supplier Phenomenex Inc Supplier No. 092007
GC Liner for HP, Split, Straight 4mm ID x 78.5mm L x 6.3mm OD