Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation GC Performance Standard, 1.0 mg/mL Caffeine, 10 mL, Reference

Catalog No. 50201661 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This GC Performance Standard is used for gas chromatography system performance assessment. It contains 1.0 mg/mL caffeine ±1% standard diluted in 100% HPLC Grade Methylene Chloride, supplied in a 10 mL ampule.

  • Standards used for GC performance assessment
  • Contains 1.0 mg/mL caffeine ±1%
  • Diluted in 100% HPLC Grade Methylene Chloride
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: GC
  • Instrument type: GC
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Volume: 10 mL
  • Type: Reference
  • Application: System performance
  • Units per package: 1 pk
  • Hazardous material: Requires special freight handling and additional charges may apply

Catalog No. 50-201-661 Supplier Waters Corp Supplier No. 700001892
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This GC Performance Standard is used for gas chromatography system performance assessment. It contains 1.0 mg/mL caffeine ±1% standard diluted in 100% HPLC Grade Methylene Chloride, supplied in a 10 mL ampule.

  • Standards used for GC performance assessment
  • Contains 1.0 mg/mL caffeine ±1%
  • Diluted in 100% HPLC Grade Methylene Chloride
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: GC
  • Instrument type: GC
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Volume: 10 mL
  • Type: Reference
  • Application: System performance
  • Units per package: 1 pk
  • Hazardous material: Requires special freight handling and additional charges may apply

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.