Learn More
Waters Corp Waters Corporation GC Qualification Standards Kit, System Assessment, Liquid, 10 mL
Catalog No. 50201690 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Gas Chromatograph standards kit is designed for system assessment. It includes 10 ampules (10 mL each of standard solutions). All standards are prepared v/v in Iso-Octane, except for the Lindane solution. The kit also includes a Certificate of Analysis.
  • Contains 10 ampules (10 mL each) of standard solutions.
  • Includes Blank (Iso-Octane), 1-Octanol solutions at 1, 2, 4, 6, 8, and 10 μL/mL, 1% Dodecane, and 0.035 ng/μL Lindane.
  • Standards are prepared v/v in Iso-Octane (except for the Lindane solution).
  • Designed for system assessment and performance in Gas Chromatography.
  • Supports FID, TCD, and MS detection methods.
  • Liquid formulation, stored at room temperature.
  • Has a shelf life of 12 months.
  • Ships under ambient conditions.
  • Includes a certificate of analysis.
  • Contains hazardous material, which requires special freight handling.
Catalog No. 50-201-690 Supplier Waters Corp Supplier No. 700005094
This Gas Chromatograph standards kit is designed for system assessment. It includes 10 ampules (10 mL each of standard solutions). All standards are prepared v/v in Iso-Octane, except for the Lindane solution. The kit also includes a Certificate of Analysis.
  • Contains 10 ampules (10 mL each) of standard solutions.
  • Includes Blank (Iso-Octane), 1-Octanol solutions at 1, 2, 4, 6, 8, and 10 μL/mL, 1% Dodecane, and 0.035 ng/μL Lindane.
  • Standards are prepared v/v in Iso-Octane (except for the Lindane solution).
  • Designed for system assessment and performance in Gas Chromatography.
  • Supports FID, TCD, and MS detection methods.
  • Liquid formulation, stored at room temperature.
  • Has a shelf life of 12 months.
  • Ships under ambient conditions.
  • Includes a certificate of analysis.
  • Contains hazardous material, which requires special freight handling.