Learn More
Sigma Aldrich Fine Chemicals Biosciences GC-RICH PCR System dNTPack
Catalog No. 502906180
Encompass
The GC-RICH PCR System dNTPack is composed of a special enzyme blend of thermostable Taq DNA Polymerase and Tgo DNA Polymerase a thermostable enzyme with a proofreading (3-5 exonuclease) activity. This polymerase mixture by itself outperforms Taq DNA Polymerase in respect to yields fidelity and specificity beside the possibility to amplify fragments up to 5 kb in length. The GC-RICH PCR reaction buffer in combination with the included GC-RICH reSOLUTIONon SOLUTIONon allows to amplify very efficiently difficult templates like GC-rich targets.
Catalog No. 50-290-6180 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4743784001
The GC-RICH PCR System dNTPack is composed of a special enzyme blend of thermostable Taq DNA Polymerase and Tgo DNA Polymerase a thermostable enzyme with a proofreading (3-5 exonuclease) activity. This polymerase mixture by itself outperforms Taq DNA Polymerase in respect to yields fidelity and specificity beside the possibility to amplify fragments up to 5 kb in length. The GC-RICH PCR reaction buffer in combination with the included GC-RICH reSOLUTIONon SOLUTIONon allows to amplify very efficiently difficult templates like GC-rich targets.