Learn More
Enzo Life Sciences Guanylyl cyclase (soluble) (bovine lung) (5µg)
SDP
Supplier:  Enzo Life Sciences ALX202039C005
Encompass_Preferred
Alternative name: GTP pyrophosphate lyase (cycling). Concentration:
. Formulation: Liquid. In ~20µl containing 50% glycerol, 88mM NaCl, 1mM DTT, 0.5mM EDTA, 25mM triethanolamine at pH 7.4. Source: Isolated from bovine lung by an immunoaffinity method. Long Term Storage: -80°C.
Catalog No. 50-200-8046