Learn More
Trajan Scientific CTC Analytics PAL System 95 um PAL GC SPME Fiber, Carbon WR/PDMS Blue CTC, 1 Pack
Catalog No. 501019101
Encompass
CTC Analytics PAL System CTC PAL SPME Fiber 95 um Carbon WR/PDMS Hub Color-Blue, Fused Silica, Needle 23 ga, Fiber Length 10mm, Polar/Nonpolar Bi-polar, SUPELCO Equivalent Part # 57319, For Use With the PAL AutoSampler, Set of 1
Catalog No. 50-101-9101 Supplier Trajan Scientific Supplier No. FIBCWR95/10P1
CTC Analytics PAL System CTC PAL SPME Fiber 95 um Carbon WR/PDMS Hub Color-Blue, Fused Silica, Needle 23 ga, Fiber Length 10mm, Polar/Nonpolar Bi-polar, SUPELCO Equivalent Part # 57319, For Use With the PAL AutoSampler, Set of 1