Learn More
Trajan Scientific CTC Analytics PAL System 100 um, PAL GC SPME Fiber, Polydimethylsiloxane(PDMS), Red, 5 Pack
Catalog No. 501019106
Encompass
CTC Analytics PAL System CTC PAL SPME Fiber 100 um Polydimethylsiloxane(PDMS) Hub Color-Red, Fused Silica, Needle 23 ga, Fiber Length 10mm, Polar/Nonpolar NP(Nonpolar), SUPELCO Equivalent Part # 57301, For Use With the PAL AutoSampler, Set of 5
Catalog No. 50-101-9106 Supplier Trajan Scientific Supplier No. FIBP100/10P5
CTC Analytics PAL System CTC PAL SPME Fiber 100 um Polydimethylsiloxane(PDMS) Hub Color-Red, Fused Silica, Needle 23 ga, Fiber Length 10mm, Polar/Nonpolar NP(Nonpolar), SUPELCO Equivalent Part # 57301, For Use With the PAL AutoSampler, Set of 5