Learn More
Perkin Elmer US LLC Biodiesel ASTM D-6584 Standards Kit
Catalog No. 509049762
Encompass_Preferred
Standard solution 1 in 1 mL PyridineStandard solution 2 in 1 mL PyridineStandard solution 3 in 1 mL PyridineStandard solution 4 in 1 mL PyridineStandard solution 5 in 1 mL PyridineButanetriol Internal Standard in pyridine, 5 mLTricaprin Internal Standard in pyridine, 5 mL
Catalog No. 50-904-9762 Supplier Perkin Elmer US LLC Supplier No. N9331040
Standard solution 1 in 1 mL PyridineStandard solution 2 in 1 mL PyridineStandard solution 3 in 1 mL PyridineStandard solution 4 in 1 mL PyridineStandard solution 5 in 1 mL PyridineButanetriol Internal Standard in pyridine, 5 mLTricaprin Internal Standard in pyridine, 5 mL