Learn More
Perkin Elmer US LLC GC Startup Kit - 3 Gases, 0.125 in
Catalog No. 502104720
Encompass_Preferred
Includes the following pieces: (1) 50 ft x 1/8 inch Copper Tubing (3) 1/8 inch Brass Two-Piece Ferrule, Compression Tees (3) Brass 1/8 inch x 1/8 inch x 1/4 inch NPT Tees (3) Brass Adjustable Safety Relief Valves, 50 psi-150 psi (3) 1/4 inch NPT x 1/8 inch Brass Compression Fittings (3) 1/8 inch Brass Port Connectors (3) 1/8 inch Brass Ferrules (3) 1/8 inch Brass Compression Nuts (3) 1/8 inch Brass Compression Fitting Caps Tubing Cutter, 1/8 inch Tubing PTFE Tape
Catalog No. 50-210-4720 Supplier Perkin Elmer US LLC Supplier No. N9306304
Includes the following pieces: (1) 50 ft x 1/8 inch Copper Tubing (3) 1/8 inch Brass Two-Piece Ferrule, Compression Tees (3) Brass 1/8 inch x 1/8 inch x 1/4 inch NPT Tees (3) Brass Adjustable Safety Relief Valves, 50 psi-150 psi (3) 1/4 inch NPT x 1/8 inch Brass Compression Fittings (3) 1/8 inch Brass Port Connectors (3) 1/8 inch Brass Ferrules (3) 1/8 inch Brass Compression Nuts (3) 1/8 inch Brass Compression Fitting Caps Tubing Cutter, 1/8 inch Tubing PTFE Tape