Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, DC-710

Catalog No. 111004078 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4078 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4078 Supplier MilliporeSigma™ Supelco™ Supplier No. 85427100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Refractive Index: n20/D 1.537

  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
  • Density: ∼500 mPa.s, neat (25°C)
TRUSTED_SUSTAINABILITY

Specifications

Stationary Phase DC-710
Packaging Bottle
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.