Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, DC QF-1 (FS 1265)

Catalog No. 111013083 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3083 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3083 Supplier MilliporeSigma™ Supelco™ Supplier No. 21098U
Add to Cart
Add to Cart
  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
TRUSTED_SUSTAINABILITY

Specifications

Max. Temperature 225°C
Stationary Phase DC QF-1 (FS 1265)
Packaging Bottle
Quantity 50 g
Description Acetone: Soluble
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.