Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, OV-1

Catalog No. 111003897 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3897 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3897 Supplier MilliporeSigma™ Supelco™ Supplier No. 21104
Add to Cart
Add to Cart

Suitable for the separation of drugs, alkaloids, phenols, and steroids

  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
TRUSTED_SUSTAINABILITY

Specifications

Max. Temperature 350°C
Stationary Phase OV-1
Packaging Bottle
Quantity 10 g
Description Chloroform, Toluene: Soluble
For Use With (Application) Suitable for G2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.