Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, OV-101

Catalog No. 111003458 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3458 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3458 Supplier MilliporeSigma™ Supelco™ Supplier No. 21228
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A modified silicone polymer with good temperature stability, suitable for use as liquid phases in chromatography

TRUSTED_SUSTAINABILITY

Specifications

Max. Temperature 350°C
Stationary Phase OV-101
Packaging Bottle
Quantity 20 g
Description Chloroform: Soluble
For Use With (Application) Suitable for G1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.