Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, OV-17

Catalog No. 111003188 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3188 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3188 Supplier MilliporeSigma™ Supelco™ Supplier No. 21105

missing translation for 'validMainframeMsg'

Suitable for the separation of drugs, amino acid derivatives, pesticides, phenols, and sugar derivatives

  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
TRUSTED_SUSTAINABILITY

Specifications

Max. Temperature 375°C
Stationary Phase OV-17
Packaging Bottle
Quantity 25 g
Description Chloroform: Soluble
For Use With (Application) Suitable for G3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.