Learn More
MilliporeSigma™ Supelco™ GC Stationary Phase, Synperonic PE/F68
Catalog No. 111003806 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
Catalog No. Quantity
11-100-3806 50 g
1 options
Catalog No. 11-100-3806 Supplier MilliporeSigma™ Supelco™ Supplier No. 8111250G
A block copolymer of polyethylene and polypropylene glycol, mostly used as surfactant
  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
TRUSTED_SUSTAINABILITY
Specifications
Stationary Phase Synperonic PE/F68
Packaging Bottle
Quantity 50 g
For Use With (Application) Gas Chromatography (GC)