Learn More
MilliporeSigma™ Supelco™ GC Stationary Phase, Triton X-100
Catalog No. 111022446 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
Catalog No. Quantity
11-102-2446 50 g
1 options
Catalog No. 11-102-2446 Supplier MilliporeSigma™ Supelco™ Supplier No. 21123
Add to Cart
Edge
Add to Cart
Triton X-100 is a non-ionic alkylphenol ethoxylate surfactant. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time.
  • GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC)
  • GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces
  • GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients
  • Stationary phases should be stable for the chemical and thermal changes
  • Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered before choosing a stationary phase
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 g
Packaging Bottle