Learn More
Perkin Elmer US LLC GC Syringe, 10µL Fixed Needle, Point Style #2, 7cm needle
Supplier: Perkin Elmer US LLC 00230523
General-purpose point style designed for septum penetration in all chromatographic techniques. The needle has a 22° bevel with the tip slightly bent to minimize coring and needle plugging.