Learn More
Perkin Elmer US LLC GC Syringe (Fixed Needle): Capacity 100 µL, Needle Gauge 22, Point Style #3 (1/Pack)
Catalog No. 502113765
Encompass_Preferred
Each syringe is individually inspected for accuracy and performance. Needle Gauge: 22 Tip Description: LC
Catalog No. 50-211-3765 Supplier Perkin Elmer US LLC Supplier No. 09904822
Each syringe is individually inspected for accuracy and performance. Needle Gauge: 22 Tip Description: LC