Learn More
Perkin Elmer US LLC GC Ultramicro Volume Syringe: Capacity 1.0 µL, Needle Gauge 26 (1/Pack)
Catalog No. 502112033
Encompass_Preferred
The correct syringe is critical to ensure proper fit and operation within your instrument. Syringes must ensure a tight and secure fit in the injection port and withstand repeated injection cycles without bending or breaking. It is critical for syringes to deliver accurate and precise volumes with no dead volume or carryover between injections. Each syringe is individually inspected for accuracy and performance. Needle Gauge: 26 Tip Description: Cone
Catalog No. 50-211-2033 Supplier Perkin Elmer US LLC Supplier No. 00230111
The correct syringe is critical to ensure proper fit and operation within your instrument. Syringes must ensure a tight and secure fit in the injection port and withstand repeated injection cycles without bending or breaking. It is critical for syringes to deliver accurate and precise volumes with no dead volume or carryover between injections. Each syringe is individually inspected for accuracy and performance. Needle Gauge: 26 Tip Description: Cone