Learn More
Perkin Elmer US LLC GC Valve Starter Kit
Catalog No. 502104021
Encompass_Preferred
Includes the following pieces: (6) 2u Screen Filters, for use with Replacement Filters for N9302879 (N9302880) (1) Filter Union, 1/16 inch with Removable Screen - for use with Gas Line Filter (N9302879) (4) 1/8 inch Swagelok to 1/16 inch Valco Reducers for Packed Column Adapters (N9302917) (2) 1/8 inch Swagelok Male Nut to 1/16 inch Valvo Reducer for Injector/Detector Adapter (N9302844) (2) Stainless Steel Tubing - 50 cm x 1/16 inch O.D. x 0.02 inch I.D. for Carrier to Detector (N9302922) (2) Stainless Steel Tubing - 125 cm x 1/16 inch O.D. x 0.02 inch I.D. for Sample Input/Output (N9300087) (10) 1/16 inch Valco Ferrules (N9302835) (6) 1/16 inch Valco Nuts (N9302832)
Catalog No. 50-210-4021 Supplier Perkin Elmer US LLC Supplier No. N9302821
Includes the following pieces: (6) 2u Screen Filters, for use with Replacement Filters for N9302879 (N9302880) (1) Filter Union, 1/16 inch with Removable Screen - for use with Gas Line Filter (N9302879) (4) 1/8 inch Swagelok to 1/16 inch Valco Reducers for Packed Column Adapters (N9302917) (2) 1/8 inch Swagelok Male Nut to 1/16 inch Valvo Reducer for Injector/Detector Adapter (N9302844) (2) Stainless Steel Tubing - 50 cm x 1/16 inch O.D. x 0.02 inch I.D. for Carrier to Detector (N9302922) (2) Stainless Steel Tubing - 125 cm x 1/16 inch O.D. x 0.02 inch I.D. for Sample Input/Output (N9300087) (10) 1/16 inch Valco Ferrules (N9302835) (6) 1/16 inch Valco Nuts (N9302832)