Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GC7 sulfate | 150417-90-6 | 98.0% | 270.35 | C8H22N4O4S | 25 MG
SDP

Catalog No. 5000244081
Encompass_Preferred

GC7 sulfate is a small-molecule inhibitor of deoxyhypusine synthase (DHPS) used in biochemical and cellular research to study hypusination and translation regulation. It is supplied as a high-purity solid suitable for laboratory assays and solution preparation.

  • Inhibits deoxyhypusine synthase in biochemical and cellular assays.
  • High purity (98.0%) for consistent experimental results.
  • Solid, white to off-white appearance for easy handling and storage.
  • Molecular weight 270.35; chemical formula C8H22N4O4S for precise formulation.
  • Soluble in water (3 mg/mL); may require ultrasonic treatment and warming.
  • Store solid at 4°C in sealed, dry conditions to preserve stability.
  • Available in small research pack sizes suitable for assay development and pilot studies.

Catalog No. 50-002-44081 Supplier Medchemexpress LLC Supplier No. HY108314A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GC7 sulfate is a small-molecule inhibitor of deoxyhypusine synthase (DHPS) used in biochemical and cellular research to study hypusination and translation regulation. It is supplied as a high-purity solid suitable for laboratory assays and solution preparation.

  • Inhibits deoxyhypusine synthase in biochemical and cellular assays.
  • High purity (98.0%) for consistent experimental results.
  • Solid, white to off-white appearance for easy handling and storage.
  • Molecular weight 270.35; chemical formula C8H22N4O4S for precise formulation.
  • Soluble in water (3 mg/mL); may require ultrasonic treatment and warming.
  • Store solid at 4°C in sealed, dry conditions to preserve stability.
  • Available in small research pack sizes suitable for assay development and pilot studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.