Learn More
Medchemexpress LLC GC7 sulfate | 150417-90-6 | MFCD31619261 | 98.0% | 270.35 | C8H22N4O4S | 50 MG
SDP
Catalog No. 5000241904
Encompass_Preferred
GC7 sulfate is the sulfate salt form of GC7, a selective deoxyhypusine synthase (DHPS) inhibitor provided for laboratory research. It is a white to off-white solid, typically ≥98.0% purity, used to probe hypusination-dependent translation and cell-cycle pathways. For research use only.
  • Selective deoxyhypusine synthase inhibitor for biochemical and cellular studies.
  • Sulfate salt form with improved solubility in aqueous media.
  • High purity (98.0%) suitable for research applications.
  • White to off-white solid for easy handling and storage.
  • Available in small laboratory package sizes for experimental work.
Catalog No. 50-002-41904 Supplier Medchemexpress LLC Supplier No. HY108314A50MG
GC7 sulfate is the sulfate salt form of GC7, a selective deoxyhypusine synthase (DHPS) inhibitor provided for laboratory research. It is a white to off-white solid, typically ≥98.0% purity, used to probe hypusination-dependent translation and cell-cycle pathways. For research use only.
  • Selective deoxyhypusine synthase inhibitor for biochemical and cellular studies.
  • Sulfate salt form with improved solubility in aqueous media.
  • High purity (98.0%) suitable for research applications.
  • White to off-white solid for easy handling and storage.
  • Available in small laboratory package sizes for experimental work.