Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GC7 sulfate | 150417-90-6 | MFCD31619261 | 98.0% | 270.35 g/mol | C8H22N4O4S | 10 MG
SDP

Catalog No. 5000241970
Encompass_Preferred

GC7 sulfate is a small-molecule research reagent that inhibits deoxyhypusine synthase (DHPS). It is intended for laboratory research to probe hypusination, translation regulation, and cell-cycle pathways. The compound is supplied as a solid and as an aqueous solution, with documented solubility and storage recommendations to support biochemical and cellular assays.

  • Inhibits deoxyhypusine synthase (DHPS) in vitro and in cell assays.
  • High purity (98.0%).
  • Available as solid and as a 10 mM aqueous solution.
  • Soluble in water and PBS with ultrasonic warming and heat as needed.
  • Recommended storage: sealed at 4°C; in solvent store at -80°C for long term.
  • Suitable for biochemical and cellular research applications.

Catalog No. 50-002-41970 Supplier Medchemexpress LLC Supplier No. HY108314A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GC7 sulfate is a small-molecule research reagent that inhibits deoxyhypusine synthase (DHPS). It is intended for laboratory research to probe hypusination, translation regulation, and cell-cycle pathways. The compound is supplied as a solid and as an aqueous solution, with documented solubility and storage recommendations to support biochemical and cellular assays.

  • Inhibits deoxyhypusine synthase (DHPS) in vitro and in cell assays.
  • High purity (98.0%).
  • Available as solid and as a 10 mM aqueous solution.
  • Soluble in water and PBS with ultrasonic warming and heat as needed.
  • Recommended storage: sealed at 4°C; in solvent store at -80°C for long term.
  • Suitable for biochemical and cellular research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.