Learn More
TARGETMOL CHEMICALS INC GC7 SULFATE 25MG
SDP
Catalog No. 502581537
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GC7 Sulfate is a potent inhibitor of deoxyhypusine synthase (DHS). Eukaryotic translation initiation factor 5A2 (eIF5A2) is the only known substrate for DHS so GC7 inhibits the activation of eIF5A2 by inhibiting DHS activity. purity: 99%
Catalog No. 50-258-1537 Supplier TARGETMOL CHEMICALS INC Supplier No. T1137225MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GC7 Sulfate is a potent inhibitor of deoxyhypusine synthase (DHS). Eukaryotic translation initiation factor 5A2 (eIF5A2) is the only known substrate for DHS so GC7 inhibits the activation of eIF5A2 by inhibiting DHS activity. purity: 99%