Learn More
Selleck Chemical LLC GC7 Sulfate S2961-5mg
SDP
Catalog No. 503111693
Encompass_Preferred
GC7 (N1-guanyl-1 7-diaminoheptane) Sulfate is an inhibitor of deoxyhypusine synthase (DHPS) GC7 inhibits Neuroblastoma (NB) cell proliferation in a dose-dependent manner through induction of the cell cycle inhibitor p21 and reduction of total and phosphorylated Rb proteins
Catalog No. 50-311-1693 Supplier Selleck Chemical LLC Supplier No. S29615MG
Add to Cart
Add to Cart
GC7 (N1-guanyl-1 7-diaminoheptane) Sulfate is an inhibitor of deoxyhypusine synthase (DHPS) GC7 inhibits Neuroblastoma (NB) cell proliferation in a dose-dependent manner through induction of the cell cycle inhibitor p21 and reduction of total and phosphorylated Rb proteins