Learn More
Aviva Systems Biology Corp Gcat Antibody - C-terminal region (ARP48753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gcat. It was validated on Western Blot. . Alternate names: AI526977, KblApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-377