Learn More
Enzo Life Sciences GCDFP-15 (human) monoclonal antibody (D6) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX801051L001
Encompass_Preferred
Clone: D6. Isotype: IgG2a. Immunogen: GCDFP-15 (gross cystic disease fluid protein-15; BRST-2). Species reactivity: Human. Applications: IHC (PS). Formulation: Liquid. In 0.01M PBS containing 1% BSA and 0.1% sodium azide.
Catalog No. 50-200-9712