Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5304MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: PIP/1571. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HepG2 Cells. Breast or Pancreas. Cellular Staining: Cytoplasmic (Golgi bodies) and Secreted. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human GCDFP-15 protein fragment (aa 41-146). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 15kDa.Chromosome: 7q34. Gene Symbol: PIP. Gene ID: 5304. Ensemble: ENSG00000159763. Uniprot: NP_002643.
Catalog No. 50-188-5055